missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD196 (CCR6) Monoclonal Antibody (R6H1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-1969-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development.Specifications
CD196 | |
Monoclonal | |
5μL (0.125μg)/test | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
25 Tests | |
Primary | |
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures. |
Flow Cytometry | |
R6H1 | |
PE-Cyanine7 | |
Mouse | |
RUO | |
Human | |
IgG1 |
For Research Use Only.