missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD196 (CCR6) Monoclonal Antibody (R6H1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-1969-42
Code : Z2
Additional Details : Weight : 0.50000kg
Description
CD196 (CCR6) Monoclonal antibody specifically detects CD196 (CCR6) in Human samples. It is validated for Flow Cytometry.Specifications
CD196 (CCR6) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P51684 | |
CCR6 | |
Affinity chromatography | |
RUO | |
1235 | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
Flow Cytometry | |
R6H1 | |
PE-Cyanine7 | |
CCR6 | |
BN-1; CC chemokine receptor 6; C-C chemokine receptor type 6; C-C CKR-6; C-C motif chemokine receptor 6; CC-CKR-6; CCR6; CCR-6; CD196; chemokine (C-C motif) receptor 6; chemokine (C-C) receptor 6; chemokine receptor-like 3; CKR6; CKRL3; CKR-L3; Cmkbr6; DCR2; DRY6; DRY-6; G protein-coupled receptor 29; GPR29; GPRCY4; GPR-CY4; G-protein coupled receptor 29; KY411; LARC receptor; seven-transmembrane receptor, lymphocyte, 22; STRL22 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Liquid |
For Research Use Only.