missing translation for 'onlineSavingsMsg'
Learn More

CD196 (CCR6), PE, clone: R6H1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1969-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556936

  • £80.99 / 25 tests


EU Stock 1
Add to basket

Description

Description

This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development.
Specifications

Specifications

CD196
R6H1
PE
Conjugated
IgG1
RUO
Primary
Human
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.