missing translation for 'onlineSavingsMsg'
Learn More

CD196 (CCR6) Mouse anti-Human, FITC, Clone: R6H1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1969-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596506

  • £165.00 / 100 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors. TRAIL/Apo2L is a new member of the TNF family and induces apoptosis of a variety of tumor cell lines. DR4 and DR5 are the recently identified functional receptors for TRAIL, and DcR1/TRID is a decoy receptor. Another member of the TRAIL receptor family was more recently identified and designated DcR2, TRAIL-R4, or TRUNDD. DcR2 has an extracellular TRAIL-binding domain but lacks intracellular death domain and does not induce apoptosis. Like DR4 and DR5, DcR2 transcript is widely expressed in normal human tissues. Overexpression of DcR2 attenuated TRAIL-induced apoptosis.
Specifications

Specifications

CD196 (CCR6)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P51684
BN-1, CKR-L3, CKR6, CKRL3, CMKBR6, DCR2, DRY-6, GPR-CY4, GPR29, GPRCY4, STRL22
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
R6H1
FITC
CCR6
Liquid
CCR6
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1235
Videos
SDS
Documents

For Research Use Only.