Learn More
CD195 (CCR5) Mouse anti-Human, PE-Cyanine7, Clone: NP-6G4, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 25-1956-41
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15568506
  • £85.00 / 25 tests
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120.
Specifications
Specifications
CD195 (CCR5)
NP-6G4
PE-Cyanine7
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
C-C chemokine receptor type 5, CMKBR5
IgG1, kappa