missing translation for 'onlineSavingsMsg'
Learn More

CD195 (CCR5), PE, clone: NP-6G4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1956-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15526936

  • £74.97 / 25 tests


EU Stock 2
Add to basket

Description

Description

Description: The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120.Applications Reported: This NP-6G4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This NP-6G4 antibody has been pre-titrated and tested on normal human peripheral blood cells. For optimal performance, it is recommended that staining with NP-6G4 be done at 37°C for 1 hour in Flow Cytometry Staining Buffer (cat. 00-4222). This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD195 (CCR5)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P51681
C-C CKR-5, CC-CKR-5, HIV-1 fusion coreceptor, CHEMR13, CD195 antigen, CMKBR5
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
NP-6G4
PE
CCR5
Conjugated
CCR5
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1234
Videos

For Research Use Only.