missing translation for 'onlineSavingsMsg'
Learn More

CD195 (CCR5), PE, clone: eBioT21/8 (T21/8), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1957-41

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15290304

  • £68.15 / 25 tests


Estimated Shipment date
from Supplier 27-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioT21/8 monoclonal antibody reacts with the amino terminus of human CCR5 (CD195), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1alpha, MIP-1beta and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120. Binding of the eBioT21/8 monoclonal antibody displays donor variability and may be affected by sub-optimal staining conditions.Applications Reported: This eBioT21/8 (T21/8) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioT21/8 (T21/8) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. For optimal performance, it is recommended that staining with eBioT21/8 be done at room temperature or 37°C for 1 hour in Flow Cytometry Staining Buffer (cat. 00-4222). This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD195 (CCR5)
eBioT21/8 (T21/8)
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CCR-5
IgG1
RUO
Primary
Human
Videos