CD195 (CCR5) Mouse anti-Human, APC, Clone: NP-6G4, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 17-1956-41

25TEST Anti-Human CD195 (CCR5) APC

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15548046

Quantity Price
1 £ 78.88 / 25 tests
Estimated Shipment date
from Supplier 01-03-2017
Add to basket


Description and Specification

Specification

Antigen CD195 (CCR5)
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias C-C chemokine receptor type 5, CMKBR5
Host Species Mouse
Applications Flow Cytometry
Isotype IgG1
Quantity 25 tests
Regulatory Status RUO
Species Reactivity Human
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone NP-6G4
Concentration 5μL (0.25μg)/test
Conjugate APC

The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120.