missing translation for 'onlineSavingsMsg'
Learn More

CD195 (CCR5), PE, clone: HM-CCR5 (7A4), eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1951-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228709

  • £68.46 / 50µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: CCR5 is the major co-receptor for macrophage-tropic isolates of HIV-1. CCR5, also known as CD195, is a seven transmembrane chemokine receptor capable of binding RANTES, MIP-1 alpha and MIP-1 beta. Knockout mice show a decreased recruitment of macrophages and increased susceptibility to infection. Protein and mRNA has been found in macrophages from thioglycolate-treated and activated T cells. The HM-CCR5(7A4) monoclonal antibody binds the N terminal extracellular domain of mouse CCR5 with no crossreactivity to human CCR5.Applications Reported: This HM-CCR5 (7A4) antibody has been reported for use in flow cytometric analysis.Applications Tested: This HM-CCR5 (7A4) antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal-exudate cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD195
HM-CCR5 (7A4)
PE
Conjugated
Armenian Hamster
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MIP-1 alpha Receptor C-C chemokine receptor type 5, Cmkbr5
IgG
RUO
Primary
Murine
Videos