missing translation for 'onlineSavingsMsg'
Learn More

CD193 (CCR3) Mouse anti-Human, PE, Clone: eBio5E8-G9-B4 (5E8-G9-B4), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1939-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586926

  • £52.50 / 25 tests

Estimated Shipment
26-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CCR3 (C-C chemokine receptor type 3) binds to eotaxin, eotaxin-3, MCP-3, MCP-4, RANTES, and MIP-1 delta. CCR3 trasnduces a signal by increasing the intracellular calcium ions level. It also serves as an alternative co-receptor with CD4 for HIV-1 infection. Other chemokines play important roles in inflammation and are critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines.
Specifications

Specifications

CD193 (CCR3)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P51677
b-chemokine receptor; C-C chemokine receptor type 3; C-C CKR-3; CC chemokine receptor 3; CC-CKR-3; CCR-3; chemokine (C-C motif) receptor 3; chemokine (C-C) receptor 1,-like 2; chemokine (C-C) receptor 3; CKR3; CMKBR3; eosinophil CC chemokine receptor 3; eosinophil eotaxin receptor; macrophage inflammatory protein 1-alpha receptor-like 2; MGC102841; MIP-1 alpha RL2; MIP-1 alphaRL2; probable C-C chemokine receptor type 3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio5E8-G9-B4 (5E8-G9-B4)
PE
CCR3
Liquid
CCR3
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1232
Videos
Documents

For Research Use Only.