missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, PE-Cy7, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £182.00
Specifications
Antigen | CD19 |
---|---|
Clone | 1D3 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961768
|
EnquireBio
QAB28-PE7-25UG |
25 μg |
£71.60
25µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951768
|
EnquireBio
QAB28-PE7-100UG |
100 μg |
£182.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD19 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12478 | |
IgG2a κ | |
Protein A/G Purified |
1D3 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_033974 | |
Cd19 | |
Primary | |
Store at 2-4°C. |