Learn More
CD19 Rat, APC, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB28-APC-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15941758
  • £69.32 / 25µg
Estimated Shipment
29-04-2021
Specifications
CD19
1D3
APC
NP_033974
Rat
Protein A/G Purified
RUO
Primary
12478
Flow Cytometry
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd19
IgG2a kappa
25 μg
Store at 2-4°C.