missing translation for 'onlineSavingsMsg'
Learn More

CD19, PE-Cyanine5.5, clone: eBio1D3 (1D3), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 35-0193-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381020

  • £76.80 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.Applications Reported: This 1D3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio1D3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-567 nm; Emission: 690 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Specifications

Specifications

CD19
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P25918
AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio1D3 (1D3)
PE-Cyanine5.5
Cd19
Conjugated
Cd19
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
12478
Videos

For Research Use Only.