Learn More
CD19 Monoclonal Antibody (J3-129), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-0197-41
86.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This J3-129 monoclonal antibody reacts with human CD19, a 95-kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development, excluding terminally differentiated plasma cells. However, follicular dendritic cells express CD19. CD21, CD81, Leu13, MHC class II, and CD19 together form a multi-molecular complex that associates with the B cell receptor. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and B cell proliferation. Applications Reported: This J3-129 antibody has been reported for use in flow cytometric analysis. Applications Tested: This J3-129 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P15391 | |
Cd19 | |
Affinity chromatography | |
RUO | |
930 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
J3-129 | |
PerCP-eFluor 710 | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.