missing translation for 'onlineSavingsMsg'
Learn More

CD19, eVolve 605, clone: SJ25C1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 83-0198-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15580747

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Description: The SJ25C1 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express this molecule. CD19, along with CD21, CD81, Leu13, and MHC class II, form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.Applications Reported: This SJ25C1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This SJ25C1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 605 can be optimally excited with laser lines from 325-405 nm, and emits near 610 nm. We recommend using the violet (405 nm) laser line for excitation and a 595LP with a 605/40 or 610/20 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-590 nm; Emission: 605 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

CD19
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P15391
AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
SJ25C1
eVolve 605
CD19
Conjugated
CD19
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary
930
Videos
Documents

For Research Use Only.