missing translation for 'onlineSavingsMsg'
Learn More

CD19, clone: LC1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0190-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15206837

  • £30.83 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The LC1 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multi-molecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The antibody recognizes a sequence near the carboxy terminus.Applications Reported: This LC1 antibody has been reported for use in western blotting and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This LC1 antibody has been tested by western blot and can be used at less than or equal to 5 μg/mL. This antibody has also been tested by immunohistochemistry on FFPE human tonsil using high pH antigen retrieval at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD19
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P15391
AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
LC1
Unconjugated
CD19
Purified
CD19
IgG1, kappa
25μg
Store at 2-8°C.
Primary
930
Videos

For Research Use Only.