missing translation for 'onlineSavingsMsg'
Learn More

CD19, Alexa Fluor 700, clone: HIB19, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0199-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15520347

  • £93.52 / 25 tests


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

The HIB19 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.
Specifications

Specifications

CD19
HIB19
Alexa Fluor 700
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leu-12
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.