missing translation for 'onlineSavingsMsg'
Learn More

CD19, Alexa Fluor 700, clone: HIB19, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0199-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15530347

  • £182.82 / 100 tests


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HIB19 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.Applications Reported: This HIB19 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIB19 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD19
HIB19
Alexa Fluor 700
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leu-12
IgG1
RUO
Primary
Human
Videos