missing translation for 'onlineSavingsMsg'
Learn More

CD185 (CXCR5), PerCP-eFluor™ 710, clone: SPRCL5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-7185-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15559356

  • £228.19 / 100µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

This SPRCL5 monoclonal antibody reacts with Mouse CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt’s lymphoma. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. CXCR5+ circulating T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CXCR5 is expressed on nearly all CD4+ cells along with CD45RO and such activation markers as CD69 and ICOS. Tonsillar CXCR5+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing B cell help. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CXCR5 an established marker of follicular helper T cells.

This SPRCL5 does not see a formaldehyde fixed epitope, therefore staining should be done prior to fixation.
Specifications

Specifications

CD185 (CXCR5)
SPRCL5
PerCP-eFluor 710
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
C-X-C Chemokine Receptor Type 5, BLR1, MDR15
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.