missing translation for 'onlineSavingsMsg'
Learn More

CD184 (CXCR4) Rat anti-Mouse, eFluor® 450, Clone: 2B11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-9991-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15579856

  • £200.00 / 100µg

EU Stock 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered. Receptor for the C-X-C chemokine CXCL12/SDF-1 that transduces a signal by increasing intracellular calcium ion levels and enhancing MAPK1/MAPK3 activation. Acts as a receptor for extracellular ubiquitin; leading to enhanced intracellular calcium ions and reduced cellular cAMP levels. Involved in hematopoiesis and in cardiac ventricular septum formation. Also plays an essential role in vascularization of the gastrointestinal tract, probably by regulating vascular branching and/or remodeling processes in endothelial cells. Involved in cerebellar development. In the CNS, could mediate hippocampal-neuron survival. Acts as a coreceptor (CD4 being the primary receptor) for HIV-1 X4 isolates and as a primary receptor for some HIV-2 isolates. Promotes Env-mediated fusion of the virus. Binds bacterial lipopolysaccharide (LPS) et mediates LPS-induced inflammatory response, including TNF secretion by monocytes.
Specifications

Specifications

CD184 (CXCR4)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P70658
CXC Chemokine Receptor 4, Fusin, LESTR, HUMSTR
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
2B11
eFluor™ 450
Cxcr4
Liquid
Cxcr4
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12767
Videos
Documents

For Research Use Only.