missing translation for 'onlineSavingsMsg'
Learn More

CD184 (CXCR4), Biotin, clone: 12G5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9999-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15310160

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The 12G5 monoclonal antibody reacts with human CD184 (CXCR4), also termed Fusin, LESTR, or HUMSTR. CD184 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CD184 is expressed predominantly on naive T cell subsets of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. The 12G5 antibody shows partial inhibition of chemotaxis and calcium influx induced by SDF-1 (the natural ligand of CD184), blocks CD4-independent HIV-2 infection, and blocks CD4-dependent infection by some T-tropic HIV-1 isolates.Applications Reported: The 12G5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 12G5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD184 (CXCR4)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P61073
CXC Chemokine Receptor 4, Fusin, LESTR, HUMSTR
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
12G5
Biotin
CXCR4
Conjugated
CXCR4
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
7852
Videos

For Research Use Only.