Learn More
CD183 (CXCR3) Mouse anti-Human, PE-Cy7, Clone: CEW33D, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 25-1839-42
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15558506
  • £225.61 / 100 tests
Estimated Shipment date
from Supplier 03-07-2017
Add to basket
Disclaimers:
For Research Use Only.
Specifications
Description
The CEW33D monoclonal antibody reacts with human CD183. CD183, also known as CXCR3, is a G protein-coupled chemokine receptor that interacts with ligands CXCL9 (MIG), CXCL10 (IP-10), and CXCL11 (I-TAC). Strongly associated with type 1 immunity, CD183 is induced in naïve T cells upon activation and remains upregulated in T helper type (Th)1 cells, CD8 effector cells, NK cells and NKT cells. CD183-ligand interactions mediate infiltration of inflamed tissues in normal type 1 immune responses as well as in many inflammatory and autoimmune diseases. CD183 is also expressed on some B cells and plasmacytoid DC.
Description & Specifications
Specifications
CD183 (CXCR3)
CEW33D
PE-Cy7
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
Conjugated
C-X-C chemokine receptor type 3, CXC-R3, GPR9
IgG1