missing translation for 'onlineSavingsMsg'
Learn More

CD182 (CXCR2) Mouse anti-Human, PerCP-eFluor™ 710, Clone: eBio5E8-C7-F10 (5E8-C7-F10), eBioscience™

Mouse Monoclonal Antibody

£81.00 - £190.00

Specifications

Antigen CD182 (CXCR2)
Clone ebio5E8-C7-F10 (5E8-C7-F10)
Target Species Human
Conjugate PerCP-eFluor™ 710
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15569206 View Documents Affymetrix eBioscience
46-1829-41
25 Tests £81.00
25 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-05-2020
Please to purchase this item. Need a web account? Register with us today!
15579206 View Documents Affymetrix eBioscience
46-1829-42
100 Tests £190.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-05-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
Specifications

Specifications

CD182 (CXCR2)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
ebio5E8-C7-F10 (5E8-C7-F10)
PerCP-eFluor™ 710
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.