missing translation for 'onlineSavingsMsg'
Learn More

CD182 (CXCR2) Mouse anti-Human, PerCP-eFluor™ 710, Clone: eBio5E8-C7-F10 (5E8-C7-F10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-1829-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579206

  • £190.00 / 100 tests

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
Specifications

Specifications

CD182 (CXCR2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P25025
CXCR2, CDw128b, CD182 antigen, CMKAR2, CXCR-2, CD182, CXC-R2, IL-8R B, Il-8 receptor b, IL-8 receptor type 2, Il-8rb, IL8R2, IL8RB, IL-8 receptor beta
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
ebio5E8-C7-F10 (5E8-C7-F10)
PerCP-eFluor™ 710
CXCR2
Liquid
CXCR2
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3579
Videos
Documents

For Research Use Only.