missing translation for 'onlineSavingsMsg'
Learn More

CD180 (RP105), PE, clone: MHR73-11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1809-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516926

  • £75.29 / 25 tests


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The MHR73-11 monoclonal antibody reacts with human CD180 (RP105). This 105kDa type I transmembrane molecule is a member of the TLR family of proteins characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. RP105 physically associates with another molecule called MD-1 and is expressed on B, monocytes/macrophages, and dendritic cells. Histological studies show that RP105 is expressed mainly on mature B cells in mantle zones, while germinal center cells are either dull or negative. The RP105/MD-1 complex in concert with TLR4 mediates B cell recognition and signaling of LPS. MHR73-11 activates B cells, leading to increases in cell size, expression of the costimulatory molecule CD80, and DNA synthesis. Moreover, ligation of RP105 protects B cells from irradiation- or dexamethasone-induced apoptosis. Thus, RP105 is a signal transduction molecule and plays a role in regulation of B cell growth and death. A significant proportion of circulating B cells in SLE patients is RP105 negative. Loss of RP105 is associated with B cell activation and increased disease activity in SLE patients.Applications Reported: MHR73-11 has been reported for use in flow cytometric analysis.Applications Tested: This MHR73-11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD180 (RP105)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q99467
LY64, Ly78, MGC126233, MGC126234, RP105
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MHR73-11
PE
CD180
Conjugated
CD180
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
4064
Videos

For Research Use Only.