missing translation for 'onlineSavingsMsg'
Learn More

CD18 (LFA-1 beta), PE, clone: M18/2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0181-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15248469

  • £49.41 / 50µg


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The M18/2 monoclonal antibody reacts with mouse CD18, the 90-95kDa integrin beta 2 glycoprotein. CD18 is the common beta subunit that non-covalently associates with CD11a (alpha L), CD11b (alpha M) and CD11c (alpha X) to form LFA-1, Mac-1 and gp150/95. Heterodimers of CD18 and the a subunits are expressed broadly by all leukocytes with NK and T cells showing higher density of surface expression. Through the interactions of LFA-1, Mac-1 and gp150/95 with their counter-receptors, CD18 is involved in cellular adhesion and the immune response.Applications Reported: The M18/2 antibody has been reported for use in flow cytometric analysis.Applications Tested: The M18/2 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 1.0ug per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD18
M18/2
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin beta 2, ITGB2
IgG2a
RUO
Primary
Murine
Videos