missing translation for 'onlineSavingsMsg'
Learn More

CD18 (LFA-1 beta), clone: M18/2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0181-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15296827

  • £143.47 / 500µg


Estimated Shipment
31-07-2018
Add to basket

Description

Description

Description: The M18/2 monoclonal antibody reacts with mouse CD18, the 90-95kDa integrin beta 2 glycoprotein. CD18 is the common beta subunit that non-covalently associates with CD11a (alpha L), CD11b (alpha M) and CD11c (alpha X) to form LFA-1, Mac-1 and gp150/95. Heterodimers of CD18 and the a subunits are expressed broadly by all leukocytes with NK and T cells showing higher density of surface expression. Through the interactions of LFA-1, Mac-1 and gp150/95 with their counter-receptors, CD18 is involved in cellular adhesion and the immune response.Applications Reported: The M18/2 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining. It has also been reported in blocking of CD18 in functional studies. (Please use Functional Grade purified M18/2, cat. 16-0181, in functional assays.).Applications Tested: The M18/2 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD18
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P11835
Cd18, LAD, LCAMB, Lfa1, MF17, Integrin beta-2 subunit, ITGB2, cluster designation 18
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryþFunctional AssayþImmunohistochemistryþImmunoprecipitationþWestern Blot
M18/2
Unconjugated
Itgb2
Purified
Itgb2
IgG2a, kappa
500μg
Store at 2-8°C.
Primary
16414
Videos

For Research Use Only.