missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD178 / Fas ligand Mouse, Biotin, Clone: NOK-1, enQuireBio™
Mouse Monoclonal Antibody
£99.15 - £210.00
Specifications
Antigen | CD178 / Fas ligand |
---|---|
Clone | NOK-1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952568
|
EnquireBio
QAB85-B-25UG |
25 μg |
£99.15
25µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942568
|
EnquireBio
QAB85-B-100UG |
100 μg |
£210.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD178 / Fas ligand | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
356 | |
IgG1 κ | |
Protein A/G Purified |
NOK-1 | |
Flow Cytometry | |
Biotin | |
RUO | |
NP_001289675 | |
FASLG | |
Primary | |
Store at 2-4°C. |