missing translation for 'onlineSavingsMsg'
Learn More

CD178 (Fas Ligand), APC, clone: MFL3, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5911-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15370780

  • £138.27 / 100µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MFL3 monoclonal antibody reacts with mouse Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by mouse activated T cells. The interaction of FasL with its receptor CD95 induces Fas-mediated killing. It has been reported that the human FasL antigen is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. The degree of sensitivity for the mouse antigen to MMPs has not been reported.Applications Tested: This MFL3 antibody has been tested by flow cytometric analysis of mouse FasL-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD178
MFL3
APC
Conjugated
Armenian Hamster
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FasL, CD95L, CD95 Ligand
IgG
RUO
Primary
Murine
Videos