missing translation for 'onlineSavingsMsg'
Learn More

CD178 (Fas Ligand), PE, clone: NOK-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9919-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15537276

  • £74.93 / 25 tests


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The NOK-1 monoclonal antibody reacts with human Fas (CD95) Ligand, a 40kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by neutrophils, monocytes, and activated T cells and NK cells. The interaction of FasL with its receptor (CD95, Fas) induces Fas-mediated killing of lymphocytes. Human FasL is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26kDa soluble form. Therefore for optimal detection of surface FasL on activated peripheral blood cells, incubation of cells with an MMP inhibitor is recommended.Applications Reported: NOK-1 has been reported for use in flow cytometric analysis.Applications Tested: This NOK-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD178 (CD95 Ligand)
NOK-1
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Fas ligand, FasL
IgG1
RUO
Primary
Human
Videos