missing translation for 'onlineSavingsMsg'
Learn More

CD172a (SIRP alpha), Functional Grade, clone: P84, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-1721-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298577

  • £93.40 / 50Āµg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: This P84 monoclonal antibody reacts with mouse CD172a, also known as signal regulatory protein a (SIRPa). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, macrophages, dendritic cells, but not on T and B lymphocytes. Moreover, neurons and other tissues of the central nervous system have also been shown to express CD172a. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.This monoclonal antibody has been reported to have neutralizing activity.Applications Reported: This P84 antibody has been reported for use in flow cytometric analysis and neutralization assays.Applications Tested: This P84 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD172a
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
P97797
SHPS-1, Signal-regulatory protein alpha-1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryĆ¾Functional Assay
P84
Functional Grade
Sirpa
Purified
Sirpa
IgG1, kappa
50μg
Store at 2-8°C.
Primary
19261
Videos

For Research Use Only.