missing translation for 'onlineSavingsMsg'
Learn More

CD172a (SIRP alpha), Biotin, clone: P84, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1721-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209759

  • £68.96 / 25µg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: This P84 monoclonal antibody reacts with mouse CD172a, also known as signal regulatory protein a (SIRPa). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, macrophages, dendritic cells, but not on T and B lymphocytes. Moreover, neurons and other tissues of the central nervous system have also been shown to express CD172a. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.This monoclonal antibody has been reported to have neutralizing activity.Applications Reported: This P84 antibody has been reported for use in flow cytometric analysis.Applications Tested: This P84 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD172a
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P97797
SHPS-1, Signal-regulatory protein alpha-1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
P84
Biotin
Sirpa
Conjugated
Sirpa
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
19261
Videos

For Research Use Only.