missing translation for 'onlineSavingsMsg'
Learn More

CD172a (SIRP alpha), APC, clone: P84, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1721-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15380690

  • £87.34 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: This P84 monoclonal antibody reacts with mouse CD172a, also known as signal regulatory protein a (SIRPa). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, macrophages, dendritic cells, but not on T and B lymphocytes. Moreover, neurons and other tissues of the central nervous system have also been shown to express CD172a. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.This monoclonal antibody has been reported to have neutralizing activity.Applications Reported: This P84 antibody has been reported for use in flow cytometric analysis.Applications Tested: This P84 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD172a
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P97797
SHPS-1, Signal-regulatory protein alpha-1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
P84
APC
Sirpa
Conjugated
Sirpa
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
19261
Videos

For Research Use Only.