missing translation for 'onlineSavingsMsg'
Learn More

CD172a (SIRP alpha), FITC, clone: 15-414, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1729-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556506

  • £143.78 / 100 tests


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Description: This 15-414 monoclonal antibody reacts with human CD172a, also known as signal regulatory protein alpha (SIRP alpha). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, granulocytes, dendritic cells, and CD34+CD38- hematopoietic progenitor cells but not on T and B lymphocytes. Moreover, neurons have also been shown to express CD172. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.Applications Reported: This 15-414 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 15-414 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD172a (SIRP alpha)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P78324
SIRPa, SHPS-1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
15-414
FITC
SIRPA
Conjugated
SIRPA
IgG2a
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
140885
Videos

For Research Use Only.