missing translation for 'onlineSavingsMsg'
Learn More

CD171, APC, clone: eBio5G3 (5G3), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1719-42

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15331916

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only.


Description

Description

The monoclonal antibody eBio5G3 recognizes CD171 also known as neural cell adhesion molecule L1. CD171 is a member of the Ig superfamily containing 6 extracellular Ig domains and five fibronectin type III-like repeats. CD171 has been shown to function as a cell adhesion molecule mediating homotypic and heterotypic cell-cell interactions in neuronal myelination, neurite outgrowth and regeneration. Expression of CD171 has been found on monocytes and mature monocytic-derived and follicular DCs, a minor subset of lymphocytes in addition to that found on neuronal tissue and some tumor cells lines. Expression of CD171 on tumors is thought to contribute to tumor progression. Epitope of eBio5G3 is in amino-terminal Ig-like domain.
Specifications

Specifications

CD171
eBio5G3 (5G3)
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Neural Cell Adhesion Molecule, L1, NCAM L1
IgG2a
RUO
Primary
Human
Videos