missing translation for 'onlineSavingsMsg'
Learn More

CD169 (Siglec-1) Mouse anti-Human, PE-eFluor® 610, Clone: 7-239, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-1699-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15580467

  • £217.00 / 100 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
Specifications

Specifications

CD169 (Siglec-1)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZZ2
CD169, SIGLEC-1, SN, dJ1009E24.1, sialic acid-binding Ig-like lectin 1, sialic acid-binding immunoglobulin-like lectin 1, sialoadhesin
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
7-239
PE-eFluor™ 610
SIGLEC1
Liquid
SIGLEC1
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6614
Videos
SDS
Documents

For Research Use Only.