missing translation for 'onlineSavingsMsg'
Learn More

CD169 (Siglec-1) Mouse anti-Human, PE, Clone: 7-239, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1699-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596916

  • £78.25 / 25 tests

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
Specifications

Specifications

CD169 (Siglec-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZZ2
CD169, SIGLEC-1, SN, dJ1009E24.1, sialic acid-binding Ig-like lectin 1, sialic acid-binding immunoglobulin-like lectin 1, sialoadhesin
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
7-239
PE
SIGLEC1
Liquid
SIGLEC1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6614
Videos
Documents

For Research Use Only.