Learn More
CD166 (ALCAM) Monoclonal Antibody (eBioALC48), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 17-1661-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: Activated leukocyte cell adhesion molecule (ALCAM; CD166) is a 105 kD cell-surface adhesion molecule belonging to the Ig superfamily of receptors. ALCAM is a type I transmembrane glycoprotein, possessing five extracellular Ig domains, a transmembrane domain, and a short cytoplasmic tail. ALCAM is involved in homotypic adhesion, as well as heterotypic adhesion with its natural ligand, CD6. A soluble isoform of ALCAM has been shown to inhibit homophilic adhesive interactions. ALCAM is expressed on a variety of cell types, including activated T and B cells, monocyte-derived dendritic cells, and thymic epithelial cells. ALCAM expression has also been used as a marker for the isolation of pluripotent stem cells. Applications Reported: This eBioALC48 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioALC48 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD166 (ALCAM) Monoclonal antibody specifically detects CD166 (ALCAM) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD166 (ALCAM) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q61490 | |
ALCAM | |
Affinity chromatography | |
RUO | |
11658 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioALC48 | |
APC | |
ALCAM | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.