missing translation for 'onlineSavingsMsg'
Learn More

CD166 (ALCAM) Monoclonal Antibody (3A6), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1668-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586916

  • £220.00 / 100 tests
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This 3A6 monoclonal antibody reacts with human CD166, which is also known as activated leukocyte cell adhesion molecule (ALCAM). CD166 is an immunoglobulin superfamily cell adhesion molecule that is broadly expressed on developing and adult hematopoietic (e.g., activated T and B cells, monocytes, and dendritic cells) and non-hematopoietic (e.g., neural, endothelial, and epithelial) tissues. CD166 has also been detected in melanoma, colorectal carcinoma, as well as cancers of the breast, prostate, and bladder. CD166 mediates cell-cell adhesion by forming low-affinity homophilic interactions (ALCAM-ALCAM) or high-affinity heterophilic interactions with its ligand CD6 (ALCAM-CD6). CD166 plays a role in development, inflammation, and neutrophil migration. Moreover, its expression can be used as a prognostic marker for several cancers and has been reported to be a cancer stem cell marker. The 3A6 antibody has been reported to induce the internalization of CD166 and its recycling back to the cell surface. Applications Reported: This 3A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3A6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
Specifications

Specifications

CD166 (ALCAM)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q13740
ALCAM
Affinity chromatography
RUO
214
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
3A6
PE
ALCAM
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.