missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163, Biotin, clone: eBioGHI/61 (GHI/61), eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-1639-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
The eBioGHI/61 monoclonal antibody reacts with human CD163 (Haemoglobin Scavenger Receptor, M130), a member of the macrophage scavenger receptor family. CD163 is expressed by tissue macrophages, circulating blood monocytes, and some reports suggest that it may also be expressed by haematopoietic progenitor cells. CD163 functions as a high affinity scavenger receptor for the complex of haemoglobin and haptoglobin which forms to clear haemoglobin from the blood. The high affinity for CD163 is specific for the haemoglobin-haptoglobin complex, whereas haemoglobin or haptoglobin display no to low affinity toward CD163. Upon ligand binding, CD163 initiates signal transduction which leads to the production of IL-6 and IL-10. Furthermore, it has been demonstrated that cytokines such as IL-6 and IL-10 can induce expression of CD163, whereas proinflammatory stimuli, such as LPS or TNF alpha, can cause shedding of a soluble form of CD163, whose function is unclear.Specifications
CD163 | |
eBioGHI/61 (GHI/61) | |
Biotin | |
Conjugated | |
Murine | |
25μg | |
Store at 2-8°C. Do not freeze. | |
Monoclonal |
Flow Cytometry | |
0.5mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
M130, Haemoglobin Scavenger Receptor, Macrophage marker | |
IgG1, kappa | |
RUO | |
Primary | |
Human |
For Research Use Only.