missing translation for 'onlineSavingsMsg'
Learn More

CD162 (PSGL-1) Mouse anti-Human, PE, Clone: FLEG, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1629-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566916

  • £151.00 / 100 tests

Estimated Shipment
04-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SELPLG is the high affinity counter-receptor for P-selectin on myeloid cells and stimulated T lymphocytes. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. The organization of the SELPG gene closely resembles that of CD43 and the human platelet glycoprotein GpIb-alpha both of which have an intron in the 5-prime-noncoding region, a long second exon containing the complete coding region, and TATA-less promoters.
Specifications

Specifications

CD162 (PSGL-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q14242
CD162; CLA; cutaneous lymphocyte-associated associated antigen; P-Selectin Glycoprotein Ligand 1; PSGL1; SELPG
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
FLEG
PE
SELPLG
Liquid
SELPLG
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6404
Videos
SDS
Documents

For Research Use Only.