missing translation for 'onlineSavingsMsg'
Learn More

CD161 Mouse, V450, Clone: HP-3G10, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB60-V450-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942278

  • £133.00 / 25 tests

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD161
HP-3G10
V450
NP_002249
Mouse
Protein A/G Purified
RUO
Primary
3820
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
KLRB1
IgG1 kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents