missing translation for 'onlineSavingsMsg'
Learn More

CD161, PE, clone: HP-3G10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1619-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536916

  • £75.29 / 25 tests


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: This HP-3G10 monoclonal antibody reacts with CD161 (also known as NKR-P1A), a member of the C-type lectin superfamily. The human homologue of NK1.1 in C57BL/6 mice, CD161 is expressed on natural killer cells and is upregulated in response to IL-12. CD161 can also be detected on various T cell subsets, including memory/effector CD4+ T cells, CD8+ T cells, gamma delta TCR T cells, and a subset of CD3+ thymocytes. Finally, CD161 expression has been demonstrated on human Th17 CD4+ T cells. The function of this receptor is unclear although studies suggest a possible stimulatory role. Nevertheless, Lectin-like transcript-1 (LLT1), which is also known as osteoclast inhibitory lectin, has been identified as the ligand for CD161.Applications Reported: This HP-3G10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HP-3G10 antibody has been pre-titrated and tested by flow cytometric analysis on CD4+ human peripheral T cells cultured under Th17 polarizing conditions. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD161
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q12918
Ly55a, NKR-P1A, NKRP12, Nkrp1-a, Nkrp1a
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HP-3G10
PE
KLRB1
Conjugated
KLRB1
IgG1
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3820
Videos

For Research Use Only.