missing translation for 'onlineSavingsMsg'
Learn More

CD161 Mouse anti-Human, APC, Clone: HP-3G10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1619-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568026

  • £92.25 / 25 tests

Estimated Shipment
07-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Plays an inhibitory role on natural killer (NK) cells cytotoxicity. Activation results in specific acid sphingomyelinase/SMPD1 stimulation with subsequent marked elevation of intracellular ceramide. Activation also leads to AKT1/PKB and RPS6KA1/RSK1 kinases stimulation as well as markedly enhanced T-cell proliferation induced by anti-CD3. Acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope as well as to the N-acetyllactosamine epitope. Binds also to CLEC2D/LLT1 as a ligand and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. {ECO:0000269; PubMed:16455998, ECO:0000269; PubMed:16925668, ECO:0000269; PubMed:8077657}.
Specifications

Specifications

CD161
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q12918
Ly55a, NKR-P1A, NKRP12, Nkrp1-a, Nkrp1a
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HP-3G10
APC
KLRB1
Liquid
KLRB1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3820
Videos
Documents

For Research Use Only.