missing translation for 'onlineSavingsMsg'
Learn More

CD160, PE, clone: BY55, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1609-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516916

  • £74.49 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells.Applications Reported: This BY55 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BY55 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD160
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
O95971
NK1, NK28, CD160 antigen, CD160 transmembrane isoform, CD160-delta Ig, natural killer cell receptor BY55, natural killer cell receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BY55
PE
CD160
Conjugated
CD160
IgM
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
11126
Videos

For Research Use Only.