missing translation for 'onlineSavingsMsg'
Learn More

CD160, eFluor 660, clone: BY55, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-1609-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15579906

  • £92.25 / 25 tests

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells.Applications Reported: This BY55 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BY55 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD160
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O95971
NK1, NK28, CD160 antigen, CD160 transmembrane isoform, CD160-delta Ig, natural killer cell receptor BY55, natural killer cell receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BY55
eFluor 660
CD160
Liquid
CD160
IgM
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
11126
Videos
Documents

For Research Use Only.