Learn More
CD160 Monoclonal Antibody (BY55), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£80.50 - £247.00
Specifications
Antigen | CD160 |
---|---|
Clone | BY55 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15500207
|
Affymetrix eBioscience
53-1609-42 |
100 Tests |
£247.00
100 tests |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15590197
|
Affymetrix eBioscience
53-1609-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma/delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells. Applications Reported: This BY55 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BY55 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for Flow Cytometry.Specifications
CD160 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O95971 | |
CD160 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD160 |
BY55 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
11126 | |
IgM | |
Affinity chromatography | |
Antibody |
For Research Use Only.