missing translation for 'onlineSavingsMsg'
Learn More

CD16, PE-eFluor 610, clone: eBioCB16 (CB16), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-0168-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15520417

  • £62.06 / 25 tests


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The eBioCB16 monoclonal antibody recognizes CD16 (Fc gamma RIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80 kDa. CD16 is represented by two similar genes, CD16A (Fc gamma RIIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (Fc gamma RIIIB), which exists as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1. CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3 isotypes and minimal binding of IgG2 and IgG4. Upon IgG binding, both CD16 isoforms initiate signal transduction cascades that lead to a variety of responses including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, degranulation, and proliferation.Applications Reported: This eBioCB16 (CB16) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioCB16 (CB16) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD16
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
O75015, P08637
FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioCB16 (CB16)
PE-eFluor 610
FCGR3A
Conjugated
FCGR3A, FCGR3B
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling & experimental procedures.
Primary
2214, 2215
Videos

For Research Use Only.