missing translation for 'onlineSavingsMsg'
Learn More

CD16, Cynomolgus Monkey,, PerCP-eFluor™ 710, Clone: 3G8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0166-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579086

  • £105.00 / 25 tests

Estimated Shipment date
from Supplier 20-05-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications Reported: This 3G8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3G8 antibody has been pre-titrated and tested by flow cytometric analysis of normal human and rhesus peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms – FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.
Specifications

Specifications

CD16
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O75015, P08637
FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Rhesus Monkey
Flow Cytometry
3G8
PerCP-eFluor™ 710
FCGR3A
Liquid
FCGR3A, FCGR3B
IgG1
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
2214, 2215
Videos
Documents

For Research Use Only.