missing translation for 'onlineSavingsMsg'
Learn More

CD16/CD32, eFluor 450, clone: 93, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0161-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381690

  • £146.69 / 100µg


EU Stock 1
Add to basket

Description

Description

Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils.Applications Reported: This 93 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD16/CD32
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P08101
FCGR3, FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, FCGR2, CD32, Fc gamma Receptor II, CD32 receptor 2, Fc gamma Receptor 2, FCGR2A, FCGR2B, FCGR2C, Low affinity immunoglobulin gamma Fc region receptor II-b, CDw32, Fc-gamma RII-b, Fc-gamma-RIIb, FcRII-b, FCG2, Low affinity immunoglobulin gamma Fc region receptor II-c, Fc-gamma RII-c, Fc-gamma-RIIc, FcRII-c, FGFR2B
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
93
eFluor 450
Fcgr2b
Conjugated
Fcgr2b
IgG2a, lambda
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14130
Videos

For Research Use Only.